Order Entry
Austria
ContactUsLinkComponent
Hydroxypropyl methylcellulose (Hypromellose), 50 cP FCC
Hydroxypropyl methylcellulose (Hypromellose), 50 cP FCC
Supplier:  Spectrum Chemical
Certificates
Hydroxypropyl methylcellulose (Hypromellose), 50 cP FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Methoxy Groups):
    19.0 - 30.0%
  • Assay (Hydroxypropoxy Groups):
    3.0 - 12.0 %
  • Viscosity of a 2% Aqueous Solution @ 20D °C:
    40 - 60 cPs
  • Lead (Pb):
    max. 3 mg/kg
  • Loss On Drying:
    max. 5.0 %

Specification Test Results

Frequently Bought Together