Order Entry
Austria
ContactUsLinkComponent
Sodium pyruvate ≥99%, Reagent Grade
Sodium pyruvate ≥99%, Reagent Grade
Catalog # SPCMSO193-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Sodium pyruvate ≥99%, Reagent Grade
Catalog # SPCMSO193-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specification Test Results

About this item

 :
 :110.04 g/mol
Melting Pt:220…230 °C
Density:1,718 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:204-024-4
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586