Order Entry
Austria
ContactUsLinkComponent
Magnesium sulphate heptahydrate ≥99.0% USP, Ph. Eur., BP
Magnesium sulphate heptahydrate ≥99.0% USP, Ph. Eur., BP
Catalog # SPCMMA135-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  
Magnesium sulphate heptahydrate ≥99.0% USP, Ph. Eur., BP
Catalog # SPCMMA135-2.5KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Assay (MgSO₄; Anhydrous Basis):
    99.0 - 100.5%
  • pH of a 5% Solution @ 25°C:
    5.0 - 9.2
  • Loss On Ignition:
    48.0 - 52.0 %
  • Appearance of Solution:
    To Pass Test
  • Identification:
    To Pass Test

Specification Test Results

About this item

 :
 :246.48 g/mol
Storage Temperature:Ambient
CAS Number:10034-99-8
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785