Order Entry
Austria
ContactUsLinkComponent
tri-Sodium citrate dihydrate ≥99.0%, granulated USP
tri-Sodium citrate dihydrate ≥99.0%, granulated USP
Catalog # SPCMSO165-500GM
Supplier:  Spectrum Chemical
CAS Number:  
undefined
tri-Sodium citrate dihydrate ≥99.0%, granulated USP
Catalog # SPCMSO165-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (C₆H₅Na₃O₇):
    99.0 - 100.5%
  • Water:
    10.0 - 13.0 %
  • Alkalinity:
    To Pass Test
  • Tartrate:
    To Pass Test
  • Heavy Metals:
    max. 0.001 %

Specification Test Results

About this item

 :
 :294.1 g/mol
Storage Temperature:Ambient
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031