Bis(2-methoxyethyl)ether ≥99%, Reagent Grade
Catalog # SPCMB1099-200LTBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Assay:99% Min.
Specification Test Results
About this item
:
Merck Index:13,03187
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–64 °C
Density:0,937
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:203-924-4
UN:3271
ADR:3,III
:134.18 g/mol
:GHS02
:GHS08
CAS Number:111-96-6
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503