Order Entry
Austria
ContactUsLinkComponent
L(+)-Asparagine monohydrate ≥98.0% FCC
L(+)-Asparagine monohydrate ≥98.0% FCC
Catalog # SPCMA1373-100GM
Supplier:  Spectrum Chemical
CAS Number:  
L(+)-Asparagine monohydrate ≥98.0% FCC
Catalog # SPCMA1373-100GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.0 - 101.5%
  • Specific Rotation [A]D:
    +33.0 to +36.5
  • Loss On Drying:
    11.5 - 12.5 %
  • Lead (Pb):
    max. 5 mg/kg
  • Residue On Ignition:
    max. 0.1 %

Specification Test Results

About this item

 :
 :150.13 g/mol
Storage Temperature:Ambient
CAS Number:5794-13-8
Formula:C₄H₈N₂O₃·1H₂O
MDL Number:MFCD00151038