
Iron(II) sulphate heptahydrate ≥99.5%, granular FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:7782-63-0
:
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719
:278.02 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (FeSO₄.7H₂O):99.5 - 104.5%
- Lead (Pb):max. 2 mg/kg
- Mercury:max. 1 mg/kg
- Identification:To Pass Test
Specification Test Results
Frequently Bought Together