

Potassium hydroxide, pellets FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
UN:1813
ADR:8,II
:GHS05
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Assay (Total Alkali As KOH):85.0 - 100.5%
- Carbonate (As K₂CO₃):max. 3.5 %
- Insoluble Substances:To Pass Test
- Lead (Pb):max. 2 mg/kg
- Mercury:max. 0.1 MG/
Specification Test Results
Frequently Bought Together