Order Entry
Austria
ContactUsLinkComponent
Urea ≥99.0% FCC
Urea ≥99.0% FCC
Supplier:  Spectrum Chemical
Certificates
undefined
Urea ≥99.0% FCC
Supplier:  Spectrum Chemical
Certificates

Specification Test Results

  • Assay:
    99.0 - 100.5%
  • Alcohol-Insoluble Matter:
    max. 0.04 %
  • Chloride (Cl):
    max. 0.007 %
  • Lead (Pb):
    max. 5 mg/kg
  • Loss On Drying:
    max. 1.0 %

Specification Test Results

Related Information

Frequently Bought Together