Order Entry
Austria
ContactUsLinkComponent
 
Calcium chloride, anhydrous ≥93%, granular FCC
Catalog # SPCMCA138-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Calcium chloride, anhydrous ≥93%, granular FCC
Catalog # SPCMCA138-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    93% Min.

Specification Test Results

About this item

 :
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2,174 g/cm³ (20 °C)
 :110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
 :GHS07
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903