Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid 95.0-98.0% FCC
Sulphuric acid 95.0-98.0% FCC
Catalog # SPCMS1736-2.5LTGL
Supplier:  Spectrum Chemical
CAS Number:  
Sulphuric acid 95.0-98.0% FCC
Catalog # SPCMS1736-2.5LTGL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    95.0 - 98.0%
  • Arsenic (As):
    max. 3 mg/kg
  • Chloride (Cl):
    max. 0.005 %
  • Iron (Fe):
    max. 0.02 %
  • Lead (Pb):
    max. 5 mg/kg

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1,840
Storage Temperature:Ambient
UN:1830
ADR:8,II
 :GHS05
 :GHS06
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589