Order Entry
Austria
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0% FCC
Sodium sulphate, anhydrous ≥99.0% FCC
Supplier:  Spectrum Chemical
Certificates
undefined
Sodium sulphate, anhydrous ≥99.0% FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Dried Basis):
    99.0 - 100.5%
  • Lead (Pb):
    max. 2 mg/kg
  • Loss On Drying:
    max. 1 %
  • Selenium (Se):
    max. 0.003 %
  • Identification:
    To Pass Test

Specification Test Results

Frequently Bought Together