Order Entry
Austria
ContactUsLinkComponent
Propionic acid ≥99.5% FCC
Propionic acid ≥99.5% FCC
Catalog # SPCMP1326-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  
Propionic acid ≥99.5% FCC
Catalog # SPCMP1326-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    99.5% Min.

Specification Test Results

About this item

Synonyms: Propanoic acid
 :
 :74.08 g/mol
Merck Index:13,07917
Boiling Pt:141 °C (1013 hPa)
Melting Pt:–21 °C
Density:0,993
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:201-176-3
UN:3463
ADR:8.3,II
REACH:01-2119486971-24
 :GHS05
 :GHS07
CAS Number:79-09-4
Formula:C₃H₆O₂
MDL Number:MFCD00002756