Precellys® Evolution Touch Homogeniser
About this item
The Precellys Evolution Touch is both flexible and efficient, giving users the ability to grind, homogenise or lyse any kind of sample.
- Use six different tube sizes (from 0,3 to 50 ml) and prepare between 1 and 96 tubes at the same time
- From 4500 to 10000 rpm (increments of 100 rpm)
- Automatic tube locking system
- The software allows users to save an unlimited number of protocols
- Run up to 10 cycles
Easy to use with its new intuitive touchscreen interface, designed to be used with gloves, researchers can easily set up the Precellys Evolution Touch. The Precellys Evolution Touch puts the power of homogenisation into the hands of researchers and scientists.
Tube holders and other accessories for the Precellys Evolution are fully compatible with the Precellys Evolution Touch.
Certifications: E and CFR 47 part 15 certified.
2 Options Available Below
Product Details & Documents
The Precellys Evolution Touch is both flexible and efficient, giving users the ability to grind, homogenise or lyse any kind of sample.
- Use six different tube sizes (from 0,3 to 50 ml) and prepare between 1 and 96 tubes at the same time
- From 4500 to 10000 rpm (increments of 100 rpm)
- Automatic tube locking system
- The software allows users to save an unlimited number of protocols
- Run up to 10 cycles
Easy to use with its new intuitive touchscreen interface, designed to be used with gloves, researchers can easily set up the Precellys Evolution Touch. The Precellys Evolution Touch puts the power of homogenisation into the hands of researchers and scientists.
Tube holders and other accessories for the Precellys Evolution are fully compatible with the Precellys Evolution Touch.
Certifications: E and CFR 47 part 15 certified.
Documents
Precellys® Evolution Touch
Product Literature
Precellys® Evolution Touch Homogeniser
Application Notes
Specifications for Product Family
Specifications
- Catalog No:
- BERTP002511PEVT0A0
- 432-0454
- Model:
- Precellys® Evolution Touch
- No. of tubes:
- 8 - 96 wells, 300 µl
1 - 24 tubes, 2 ml/0,5 ml
1 - 12 tubes, 7 ml
1 - 6 tubes, 15 ml
1 - 3 tubes, 50 ml
1 - 6 metal tubes
- Speed range:
- 4500 - 10000 rpm (increments of 100 rpm)
- Timer:
- 10 cycles, up to 90 sec (increments 1 sec)
- W×D×H:
- 515×378×400 mm
- Weight:
- 27 kg (60 lbs.)
- Electrical:
- 90 - 250 VAC / 50 - 60 Hz Power: 1 kVA
Specifications
Product Family Options
Precellys Evolution Touch
Product Information
- AvailabilityPrice
- BERTP002511PEVT0A0Precellys® Evolution Touch, <70 dBSpecifications:
More
Specifications
Cat. No.BERTP002511PEVT0A0ModelPrecellys® Evolution TouchNo. of tubes8 - 96 wells, 300 µl
1 - 24 tubes, 2 ml/0,5 ml
1 - 12 tubes, 7 ml
1 - 6 tubes, 15 ml
1 - 3 tubes, 50 ml
1 - 6 metal tubesSpeed range4500 - 10000 rpm (increments of 100 rpm)Timer10 cycles, up to 90 sec (increments 1 sec)W×D×H515×378×400 mmWeight27 kg (60 lbs.)Electrical90 - 250 VAC / 50 - 60 Hz Power: 1 kVA
Combo Precellys Evolution and Cryolys
Product Information
Cryolys Evolution
Product Information
- Catalog #ForDescriptionAvailabilityPrice
- Cooling samples while using Precellys® Evolution homogeniserCryolys® Evolution, ONLY works with dry ice and it is placed on top of the Precellys Evolution.Log in for availability1 * 1 ST6 000,00 €
Homogenising kits
Product Information
- Catalog #DescriptionTypeForAvailabilityPrice
- Precellys® Ceramic kit 2,8 mm, reinforced, 50×2 ml tubes, pre-filled with ceramic beadsCK28-R/ 2 mlHard tissuesLog in for availability1 * 50 ST245,00 €
Lysing kits
Product Information
- Catalog #DescriptionTypeForAvailabilityPrice
- Precellys® Ceramic kit, 0,1 mm, 50×2 ml tubes, pre-filled with ceramic beadsCK01/2 mlMicroorganismsLog in for availability1 * 1 KIT176,00 €
Grinding kits
Product Information
- Catalog #DescriptionTypeForAvailabilityPrice
- Precellys® Ceramic kit 6,8 mm 'reinforced', 50×2 ml tubes, pre-filled with ceramic beadsCK68-R/ 2 mlHardest samplesLog in for availability1 * 1 KIT231,00 €
Empty tubes and beads
Product Information
- Catalog #ForDescriptionAvailabilityPrice
- Tough microorganismsCeramic beads, 0,1 mm, up to 500 tubesLog in for availability1 * 450 g214,00 €
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- 15 ml holder (adapter and blocking plate) for 6×15 ml lysis tubes15 ml holder (adapter and blocking plate) for 6×15 ml lysis tubes432-0308Precellys® EvolutionLog in for availability1 * 1 ST - 1 070,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Metal tube holder pack (1×metal tube blocking plate and 1×metal tube holder)Metal tube holder pack (1×metal tube blocking plate and 1×metal tube holder)BERTS002275PEVO0A0Precellys® EvolutionLog in for availability1 * 1 ST - 1 070,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Empty metal tubes set for Precellys® Evolution homogenisers, 6×7 ml metal tubes, tube holder and blocking plateEmpty metal tubes set for Precellys® Evolution homogenisers, 6×7 ml metal tubes, tube holder and blocking plate432-0306Personalised fillingLog in for availability1 * 1 ST - 2 510,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Cryolys® Evolution, ONLY works with dry ice and it is placed on top of the Precellys Evolution.Cryolys® Evolution, ONLY works with dry ice and it is placed on top of the Precellys Evolution.432-0346Cooling samples while using Precellys® Evolution homogeniserLog in for availability1 * 1 ST - 6 000,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 50 ml holder pack (1×50 ml tube holder working as blocking plate)50 ml holder pack (1×50 ml tube holder working as blocking plate)BERTS002474PEVO0A0Precellys® EvolutionLog in for availability1 * 1 ST - 1 060,00 €