Anti-C5orf22 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-15200R-A647
: Bioss
- Antibody type:Primary
- Antigen name:C5orf22
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:301-400/442
- Form:Liquid
- Antigen synonyms:CE022_HUMAN|C5orf22|UPF0489 protein C5orf22
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:50 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C5orf22
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C5orf22
Type: Primary
Antigen: C5orf22
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 301-400/442
Host: Rabbit
Isotype: IgG
Reactivity: Human