Anti-C5orf20 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-15199R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:C5orf20
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:131-244/244
- Form:Liquid
- Antigen synonyms:Dendritic cell nuclear protein 1|DCNP1|Chromosome 5 open reading frame 20|DCNP1_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:27 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C5orf20
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C5orf20 is a intronless gene is specifically expressed in dendritic cells (DCs), which are potent antigen-presenting cells involved in activating naive T cells to initiate antigen-specific immune response. The encoded protein is localized mainly in the perinucleus. One of the alleles (A/T) of this gene, that causes premature translation termination at aa 117, has been associated with an increased prevalence of major depression in humans
Type: Primary
Antigen: C5orf20
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 131-244/244
Host: Rabbit
Isotype: IgG
Reactivity: