Order Entry
Austria
ContactUsLinkComponent
 
Anti-C1orf53 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-15069R-A750
 :  Bioss
Anti-C1orf53 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-15069R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    C1orf53
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Epitope:
    81-145/145
  • Form:
    Liquid
  • Gene ID:
    388722
  • Antigen synonyms:
    Hypothetical protein LOC388722|Uncharacterized protein C1orf53|C1orf53|CA053_HUMAN|Chromosome 1 open reading frame 53
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    15 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C1orf51
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma. The C1orf53 gene product has been provisionally designated C1orf53 pending further characterization.

Type: Primary
Antigen: C1orf53
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 81-145/145
Host: Rabbit
Isotype: IgG
Reactivity: Rat