Anti-CISD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-13959R-A750
: Bioss
- Antibody type:Primary
- Antigen name:CISD1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:51-108/108
- Form:Liquid
- Gene ID:55847
- Antigen synonyms:mitoNEET|CDGSH iron-sulfur domain-containing protein 1|CISD1|C10orf70|ZCD1|MDS029
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:12 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CISD1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Plays a key role in regulating maximal capacity for electron transport and oxidative phosphorylation (By similarity). May be involved in Fe-S cluster shuttling and/or in redox reactions.
Type: Primary
Antigen: CISD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-108/108
Host: Rabbit
Isotype: IgG
Reactivity: Rat