Order Entry
Austria
ContactUsLinkComponent
Anti-FUT6Polyclonal Antibody (Alexa Fluor® 680)
Anti-FUT6Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13230R-A680
 :  Bioss
Anti-FUT6Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13230R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    FUT6
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    101-200/359
  • Form:
    Liquid
  • Gene ID:
    2528
  • Antigen synonyms:
    Fucosyltransferase VI|FUT6_HUMAN|EC=2.4.1.65|Galactoside 3 L fucosyltransferase|FucT VI|Alpha 1,3 fucosyltransferase|Fucosyltransferase 6|Fuc TVI|FCT3A
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    42 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human FUT6/FucT-VI
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. Fucosyltransferases catalyze the covalent association of fucose to different positional linkages in sugar acceptor molecules. The carbohydrate moieties generated and covalently attached to cell surfaces are necessary to ensure a surface contour that satisfies physiological roles, which are reliant on adhesion molecules such as Selectins (1-3). Hematopoietic lineages rely on Fucosyltransferases to confer a surface carbohydrate phenotype, which mediates proper cell adhesion molecule recruitment and cell trafficking (4-6).

Type: Primary
Antigen: FUT6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/359
Host: Rabbit
Isotype: IgG
Reactivity: