Order Entry
Austria
ContactUsLinkComponent
 
Anti-FPGT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-13211R-A750
 :  Bioss
Anti-FPGT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-13211R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    FPGT
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    251-350/594
  • Form:
    Liquid
  • Gene ID:
    8790
  • Antigen synonyms:
    GDP L fucose pyrophosphorylase|Fucose 1 phosphate guanyltransferase|GDP beta L fucose pyrophosphorylase|GDP-L-fucose diphosphorylase|FPGT_HUMAN|GDP-L-fucose pyrophosphorylase|FPGT|Fucose-1-phosphate guanylyltransferase|Fucose 1 phosphate guanylyltransferase|GFPP|GDP L fucose diphosphorylase
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    67 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human FPGT
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Guanylyltransferase enzymes transfer one molecule of GTP to another molecule and also function in the transfer of guanosine nucleotides to sugar molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. L-fucose is an important sugar in complex carbohydrates that is frequently found on plant and mammalian N-linked glycans. FPGT (Fucose-1-phosphate guanylyltransferase), also known as GFPP (GDP-L-fucose pyrophosphorylase), is a 594 amino acid cytoplasmic protein that catalyzes the formation of GDP-L-fucose from L-fucose-1-phosphate and GTP. FPGT functions to reutilize the L-fucose that is produced uopn glycoprotein and glycolipid turnover.

Type: Primary
Antigen: FPGT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/594
Host: Rabbit
Isotype: IgG
Reactivity: