Order Entry
Austria
ContactUsLinkComponent
Prostaglandin Screening Library III
Prostaglandin Screening Library III
Catalog # CAYM10503-50
Supplier:  Cayman Chemical
CAS Number:  
undefined
Prostaglandin Screening Library III
Catalog # CAYM10503-50
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Description:
    Prostaglandins in a 96-well matrix tube rack format

Specifications

About this item

For A and J series prostaglandin analog screening.

  • Contains >75 compounds
  • Includes the ''A- and J-series'' prostaglandins

The Prostaglandin Screening Library III contains more than 75 ''A and J-series'' of prostaglandins in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The library may also include thromboxanes, lactones, stable PGH, prostacyclin mimic, metabolites, and potent PGA and PGJ analogs. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.