Order Entry
Austria
ContactUsLinkComponent
Prostaglandin Screening Library I
Prostaglandin Screening Library I
Catalog # CAYM10501-50
Supplier:  Cayman Chemical
CAS Number:  
undefined
Prostaglandin Screening Library I
Catalog # CAYM10501-50
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    50 µl
  • Description:
    Prostaglandins and isoprostanes in a 96-well matrix tube rack format

Specifications

About this item

For screening of PGF analogs.

  • Contains >70 compounds
  • Includes the ''F-series'' prostaglandins and isoprostanes

The Prostaglandin Screening Library I contains more than 70 ''F-series'' of prostaglandins and isoprostanes in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.