Anti-RNF113A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9248R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RNF113A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:51-150/343
- Form:Liquid
- Gene ID:7737
- Antigen synonyms:R113A_HUMAN|Ring finger protein 113A|Ring finger protein 113A Ring finger protein 113A|RNF113|ZNF183|Zinc finger protein 183|RNF 113A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RNF113A
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
RNF113A is a novel gene whose function cannot directly be inferred from it's sequence analysis. Ring finger proteins have a role in signalling the destruction of a range of heterologous protein substrates. They are small zinc binding domains present within arrays of larger, functionally distinct proteins, often close to the amino or carboxyl termini. RNF113A is a ubiquitously expressed protein that contains a RING type zinc finger and a C3H1 type zinc finger.
Type: Primary
Antigen: RNF113A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/343
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat