Order Entry
Austria
ContactUsLinkComponent
Anti-Cell division control protein septin D1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Cell division control protein septin D1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-7755R-A750
 :  Bioss
Anti-Cell division control protein septin D1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-7755R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cell division control protein septin D1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    351-450/586
  • Form:
    Liquid
  • Gene ID:
    10801
  • Antigen synonyms:
    MLL septin-like fusion protein|SEPT9_HUMAN|PNUTL4|KIAA0991|Ovarian/Breast septin|MSF|Septin D1|SEPT9|MLL septin-like fusion protein MSF-A|Ovarian/breast septin alpha|MLL septin like fusion protein|SINT1|Cell division control protein septin D1|Septin-9|Septin 9|Ov/Br septin|MSF1|AF17q25
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    65 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human MSF/Cell division control protein septin D1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Filament-forming cytoskeletal GTPase (By similarity). May play a role in cytokinesis (Potential). May play a role in the internalisation of 2 intracellular microbial pathogens, Listeria monocytogenes and Shigella flexneri.Tissue specificity: Widely expressed. Isoforms are differentially expressed in testes, kidney, liver heart, spleen, brain, peripheral blood leukocytes, skeletal muscle and kidney. Specific isoforms appear to demonstrate tissue specificity. Isoform 5 is the most highly expressed in fetal tissue. Isoform 1 is detected in all tissues except the brain and thymus, while isoform 2, isoform 3, and isoform 4 are detected at low levels in approximately half of the fetal tissues.

Type: Primary
Antigen: Cell division control protein septin D1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 351-450/586
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse