Specifications
- Antibody type:Primary
- Antigen name:Cell division control protein septin D1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:351-450/586
- Form:Liquid
- Gene ID:10801
- Antigen synonyms:MLL septin-like fusion protein|SEPT9_HUMAN|PNUTL4|KIAA0991|Ovarian/Breast septin|MSF|Septin D1|SEPT9|MLL septin-like fusion protein MSF-A|Ovarian/breast septin alpha|MLL septin like fusion protein|SINT1|Cell division control protein septin D1|Septin-9|Septin 9|Ov/Br septin|MSF1|AF17q25
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:65 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MSF/Cell division control protein septin D1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Filament-forming cytoskeletal GTPase (By similarity). May play a role in cytokinesis (Potential). May play a role in the internalisation of 2 intracellular microbial pathogens, Listeria monocytogenes and Shigella flexneri.Tissue specificity: Widely expressed. Isoforms are differentially expressed in testes, kidney, liver heart, spleen, brain, peripheral blood leukocytes, skeletal muscle and kidney. Specific isoforms appear to demonstrate tissue specificity. Isoform 5 is the most highly expressed in fetal tissue. Isoform 1 is detected in all tissues except the brain and thymus, while isoform 2, isoform 3, and isoform 4 are detected at low levels in approximately half of the fetal tissues.
Type: Primary
Antigen: Cell division control protein septin D1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-450/586
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse