Order Entry
Austria
ContactUsLinkComponent
Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7557R-A750
Supplier:  Bioss
Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7557R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IKK alpha / beta
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    101-200/756
  • Form:
    Liquid
  • Antigen synonyms:
    NFKBIKA|I Kappa B kinase alpha|IKK alpha+IKK beta|IKK beta|IKKA_HUMAN|IkBKB|I Kappa B kinase beta|IKK alpha|IKKB|I kappa B kinase 1|IKK1|IKK2|IKKA|NFKBIKB|I kappa B kinase 2|IkBKA|IKKB_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    83 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human IKK alpha + IKK beta
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta). IKK alpha and IKK beta interact with each other and both are essential for NFkB activation.

Type: Primary
Antigen: IKK alpha / beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/756
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat