Anti-ITIH1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7018R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ITIH1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:195-290/911
- Form:Liquid
- Gene ID:3697
- Antigen synonyms:ITI-HC1|Inter-alpha-trypsin inhibitor heavy chain H1|Inter-alpha-trypsin inhibitor complex component III|ITIH1|SHAP|Serum-derived hyaluronan-associated protein|Inter-alpha-inhibitor heavy chain 1|Inter alpha trypsin inhibitor heavy chain H1|ITIH|ITI heavy chain H1|ITIH1_HUMAN|H1P
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:71 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ITIH1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localisation, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.Contains a potential peptide which could stimulate a broad spectrum of phagocytotic cells.
Type: Primary
Antigen: ITIH1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 195-290/911
Host: Rabbit
Isotype: IgG
Reactivity: