Anti-M Cadherin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6678R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:M Cadherin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:355-450/814
- Form:Liquid
- Gene ID:1013
- Antigen synonyms:MRD3|CDH15|CDHM|Muscle cadherin|CDH14|Cadherin-14|Cadherin-15|MCAD|M-cadherin|CDH3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:89 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human M-Cadherin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. M-cadherin is part of the myogenic program and may provide a trigger for terminal muscle differentiation.
Type: Primary
Antigen: M Cadherin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 355-450/814
Host: Rabbit
Isotype: IgG
Reactivity: Human