Order Entry
Austria
ContactUsLinkComponent
Anti-Delta 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Delta 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6044R-A750
Supplier:  Bioss
Anti-Delta 4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6044R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Delta 4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Epitope:
    551-650/685
  • Form:
    Liquid
  • Gene ID:
    54567
  • Antigen synonyms:
    Drosophila Delta homolog 4|hdelta2|DLL4|Delta4|Delta-like protein 4|UNQ1895/PRO4341
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    72 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human DLL4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Involved in the Notch signaling pathway as Notch ligand. Activates NOTCH1 and NOTCH4. Involved in angiogenesis; negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. Essential for retinal progenitor proliferation is required for suppressing rod fates in late retinal progenitors as well as for proper generation of other retinal cell types. During spinal cord neurogenesis, inhibits V2a interneuron fate.

Type: Primary
Antigen: Delta 4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-650/685
Host: Rabbit
Isotype: IgG
Reactivity: Mouse