Order Entry
Austria
ContactUsLinkComponent
ELISA Kit, Mouse S100A9 PicoKine
ELISA Kit, Mouse S100A9 PicoKine
Catalog # BSBTEK1152
Supplier:  Boster Bio
CAS Number:  
undefined
ELISA Kit, Mouse S100A9 PicoKine
Catalog # BSBTEK1152
Supplier:  Boster Bio
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse S100A9 PicoKine; ELISA kit

Specifications

About this item

Sandwich high sensitivity ELISA kit for quantitative detection of mouse S100A9.

  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts

Detect mouse S100A9 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. S100A9 tissue specificity.

96 wells/kit, with removable strips.