Anti-Aph-1b Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11640R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Aph-1b
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:51-150/257
- Form:Liquid
- Gene ID:83464
- Antigen synonyms:UNQ688/PRO1328|APH1B|Gamma-secretase subunit APH-1B|PSFL|Presenilin-stabilization factor-like|PRO1328|APH-1B|TAAV688|Aph-1beta
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:28 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Aph-1b
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Probable subunit of the gamma-secretase complex, an endoprotease complex that catalyses the intramembrane cleavage of integral proteins such as Notch receptors and APP (beta-amyloid precursor protein). It probably represents a stabilizing cofactor for the presenilin homodimer that promotes the formation of a stable complex. Probably present in a minority of gamma-secretase complexes compared to APH1A.
Type: Primary
Antigen: Aph-1b
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/257
Host: Rabbit
Isotype: IgG
Reactivity: Mouse