Anti-TXNDC11 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11439R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TXNDC11
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:601-700/985
- Form:Liquid
- Antigen synonyms:EF-hand-binding protein 1|Thioredoxin domain-containing protein 11|2810408E11Rik|AI427833|EFP1|RGD1307538|TXNDC11|TXD11_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:110 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TXNDC11
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H₂O₂ generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H₂O₂ generation.
Type: Primary
Antigen: TXNDC11
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 601-700/985
Host: Rabbit
Isotype: IgG
Reactivity: