Anti-NOX5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-10156R-A750
: Bioss
- Antibody type:Primary
- Antigen name:NOX5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:501-600/765
- Form:Liquid
- Gene ID:79400
- Antigen synonyms:NADPH oxidase EFhand calcium binding domain 5|NOX5|NOX5_HUMAN|MGC149776|MGC149777|NADPH oxidase 5
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:84 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NOX5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Calcium-dependent NADPH oxidase that generates superoxide. Also functions as a calcium-dependent proton channel and may regulate redox-dependent processes in lymphocytes and spermatozoa. May play a role in cell growth and apoptosis.
Type: Primary
Antigen: NOX5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/765
Host: Rabbit
Isotype: IgG
Reactivity: Human