Order Entry
Austria
ContactUsLinkComponent
Anti-GGT6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-GGT6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9909R-A680
 :  Bioss
Anti-GGT6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9909R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    GGT6
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    321-420/493
  • Form:
    Liquid
  • Gene ID:
    124975
  • Antigen synonyms:
    Gamma glutamyltranspeptidase 6|GGT 6|Gamma glutamyltransferase 6 homolog|GGT6_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human GGT6
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

GGT6 belongs to the gamma-glutamyltransferase family. GGT is a membrane-bound extracellular enzyme that cleaves gamma-glutamyl peptide bonds in glutathione and other peptides and transfers the gamma-glutamyl moiety to acceptors. GGT is also key to glutathione homeostasis because it provides substrates for glutathione synthesis. GGT6 has very limited amino acid similarity to GGT1 and it's enzymatic activities are currently uncharacterised. GGT assays are of current widespread clinical use to help assess tissue damage.

Type: Primary
Antigen: GGT6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-420/493
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat