Anti-CRHBP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-9906R-A680
: Bioss
- Antibody type:Primary
- Antigen name:CRHBP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:71-170/322
- Form:Liquid
- Gene ID:1393
- Antigen synonyms:CRF binding protein|Corticotropin releasing hormone binding protein|Corticotropin releasing factor binding protein|CRH BP|CRF BP|CRH-BP|CRFBP
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CRHBP
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
CRHBP is a potent stimulator of synthesis and secretion of preopiomelanocortin derived peptides. Although CRH concentrations in the human peripheral circulation are normally low, they increase throughout pregnancy and fall rapidly after parturition. Maternal plasma CRH probably originates from the placenta. Human plasma contains a CRH binding protein which inactivates CRH and which may prevent inappropriate pituitary adrenal stimulation in pregnancy.
Type: Primary
Antigen: CRHBP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 71-170/322
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat