Anti-XTP4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-9636R-A750
: Bioss
- Antibody type:Primary
- Antigen name:XTP4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:33-130/616
- Form:Liquid
- Gene ID:103742
- Antigen synonyms:Mien1|Rdx12|AI463380|1810046J19Rik|Migration and invasion enhancer 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:12 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human XTP4
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Increases cell migration by inducing filopodia formation at the leading edge of migrating cells. Plays a role in regulation of apoptosis, possibly through control of CASP3. May be involved in a redox-related process (By similarity).
Type: Primary
Antigen: XTP4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 33-130/616
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat