Anti-PRDX3/Peroxiredoxin 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1874R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:PRDX3/peroxiredoxin 3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:161-256/256
- Form:Liquid
- Gene ID:10935
- Antigen synonyms:Antioxidant protein 1|Protein MER5 homolog|AOP1|Peroxiredoxin III|AOP-1|PRDX3|SP-22|prx-III|PRO1748|MER5|Thioredoxin-dependent peroxide reductase, mitochondrial|HBC189|Peroxiredoxin-3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:21 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PRDX3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Involved in redox regulation of the cell. Protects radical-sensitive enzymes from oxidative damage by a radical-generating system. Acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol.
Type: Primary
Antigen: PRDX3/peroxiredoxin 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 161-256/256
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat