Anti-Inhibin beta A/Activin A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1774R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Inhibin beta A/Activin A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:331-426/426
- Form:Liquid
- Gene ID:3624
- Antigen synonyms:Activin beta-A chain|FRP|EDF|Erythroid differentiation protein|INHBA|Inhibin beta A chain
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:13/45 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Inhibin beta A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland. Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins.
Type: Primary
Antigen: Inhibin beta A/Activin A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-426/426
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat