Anti-AQP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-1506R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:AQP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:181-269/269
- Form:Liquid
- Gene ID:358
- Antigen synonyms:Water channel protein for red blood cells and kidney proximal tubule|Aquaporin-1|CO|Urine water channel|AQP-CHIP|AQP-1|Aquaporin-CHIP|AQP1|CHIP28
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:28 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human AQP1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.
Type: Primary
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 181-269/269
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat