Order Entry
Austria
ContactUsLinkComponent
 
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-0995R-A680
 :  Bioss
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-0995R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Adrenomedullin
  • Antigen symbol:
    ADM
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Epitope:
    1-100/185
  • Form:
    Liquid
  • Gene ID:
    133
  • Antigen synonyms:
    ADML_HUMAN.|AM|Proadrenomedullin N-20 terminal peptide|Adrenomedullin|ADM|PAMP|ProAM N-terminal 20 peptide|Contains|RecName|ProAM-N20
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    20 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human ADM
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterised by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is up-regulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.

Type: Primary
Antigen: Adrenomedullin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/185
Host: Rabbit
Isotype: IgG
Reactivity: Rat