Anti-SelenoProtein W Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-0495R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Selenoprotein W
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:13-88/88
- Form:Liquid
- Gene ID:20364
- Antigen synonyms:Selenoprotein W|selW|Sepw1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:9.5 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse Selenoprotein W
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Plays a role as a glutathione (GSH)-dependent antioxidant. May be involved in a redox-related process. May play a role in the myopathies of selenium deficiency.
Type: Primary
Antigen: Selenoprotein W
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 13-88/88
Host: Rabbit
Isotype: IgG
Reactivity: Rat