Anti-DOPA Decarboxylase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-0180R-A750
: Bioss
- Antibody type:Primary
- Antigen name:DOPA Decarboxylase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:201-300/480
- Form:Liquid
- Gene ID:1644
- Antigen synonyms:DOPA decarboxylase|DDC|Aromatic-L-amino-acid decarboxylase|AADC
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DDC
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
catalyses the decarboxylation of L-3,4-dihydroxyphenylalanine (DOPA) to dopamine, L-5-hydroxytryptophan to serotonin and L-tryptophan to tryptamine.
Type: Primary
Antigen: DOPA Decarboxylase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/480
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat