Anti-GFRA4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0168R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GFRA4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Epitope:51-150/299
- Form:Liquid
- Gene ID:64096
- Antigen synonyms:Persephin receptor|GFR-alpha-4|GDNF family receptor alpha-4|GDNF receptor alpha-4|GDNFR-alpha-4|GFRA4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:27 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GFRA4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Receptor for persephin. Mediates the GDNF-induced autophosphorylation and activation of the RET receptor. May be important in C-cell development and, in the postnatal development of the adrenal medulla.
Type: Primary
Antigen: GFRA4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/299
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse