Anti-Rab24 Ser95 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5690R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Rab24 Ser95
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:53917
- Antigen synonyms:Rab-24|RAB24_HUMAN|Rab 24|RAB24phospho S95|Ras-related protein Rab-24
- Modification:Phosphorylation (Ser95)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:23 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human Rab2 around the phosphorylation site of Ser95
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
A major contributor to cellular homeostasis is the ability of the cell to strike a balance between the formation and degradation/removal of its cellular components. This process of internal cellular turn-over is called autophagy (self-eating), and is facilitated by a pathway of around 16 interacting proteins in the human. The GTPase Rab24 is thought to be involved in the regulation of vesicular transport associated with autophagy.
Type: Primary
Antigen: Rab24 Ser95
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: