Anti-CD74 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-4298R-A750
: Bioss
- Antibody type:Primary
- Antigen name:CD74
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:201-296/296
- Form:Liquid
- Gene ID:972
- Antigen synonyms:HLADG|II|p33|HLA class II histocompatibility antigen gamma chain|CD74|Ia antigen-associated invariant chain|DHLAG|Ia-GAMMA|HLA-DR antigens-associated invariant chain
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:29 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HLA class II histocompatibility antigen gamma chain
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Plays a critical role in MHC class II antigen processing by stabilising peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to the endosomal/lysosomal system where the antigen processing and binding of antigenic peptides to MHC class II takes place. Serves as cell surface receptor for the cytokine MIF.
Type: Primary
Antigen: CD74
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-296/296
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse