Anti-PRDX1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-3875R-A680
: Bioss
- Antibody type:Primary
- Antigen name:PRDX1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:1-100/199
- Form:Liquid
- Gene ID:5052
- Antigen synonyms:MSP23|PAGA|Peroxiredoxin-1|PAG|Proliferation-associated gene protein|PRDX1|Thioredoxin-dependent peroxide reductase 2|Natural killer cell-enhancing factor A|NKEF-A|PRXI|NKEFA|PRX1|TDPX2|PAGB|Thioredoxin peroxidase 2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:22 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PRDX1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Involved in redox regulation of the cell. Reduces peroxides with reducing equivalents provided through the thioredoxin system but not from glutaredoxin. May play an important role in eliminating peroxides generated during metabolism. Might participate in the signaling cascades of growth factors and tumor necrosis factor-alpha by regulating the intracellular concentrations of H(2)O(2). Reduces an intramolecular disulfide bond in GDPD5 that gates the ability to GDPD5 to drive postmitotic motor neuron differentiation (By similarity).
Type: Primary
Antigen: PRDX1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/199
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Others