Specifications
- Antibody type:Primary
- Antigen name:CYP11A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:321-420/521
- Form:Liquid
- Gene ID:1583
- Antigen synonyms:Cytochrome P45(scc)|CYPXIA1|P45SCC|Cytochrome P45 11A1|Cholesterol side-chain cleavage enzyme, mitochondrial|CYP11A|CYP11A1|Cholesterol desmolase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53/57 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CYP11A1/P450SCC
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyse many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localises to the mitochondrial inner membrane and catalyses the conversion of cholesterol to pregnenolone, the first and rate-limiting step in the synthesis of the steroid hormones. Two transcript variants encoding different isoforms have been found for this gene. The cellular location of the smaller isoform is unclear since it lacks the mitochondrial-targeting transit peptide.
Type: Primary
Antigen: CYP11A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 321-420/521
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Bovine