- Antibody type:Primary
- Antigen name:DAPL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:1-100/107
- Form:Liquid
- Gene ID:92196
- Antigen synonyms:DAPL1_HUMAN|Early epithelial dferentiation-associated protein|RP23-151F21.4|EEDA|death associated protein-like 1|Death-associated protein-like 1|dapL1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:12 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DAPL1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
In contrast to growth factors which promote cell proliferation, FAS ligand (FAS-L) and the tumor necrosis factors (TNFs) rapidly induce apoptosis. Cellular response to FAS-L and TNF is mediated by structurally related receptors containing a conserved cytoplasmic region called the death domain?. DAPL1 (Death-associated protein-like 1), also known as EEDA (Early epithelial differentiation-associated protein), is a 107 amino acid protein that is expressed in hair follicles and is thought to function in a similar manner to DAP-1, possibly participating in the early stages of epithelial differentiation and/or apoptosis.
Type: Primary
Antigen: DAPL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/107
Host: Rabbit
Isotype: IgG
Reactivity: