Order Entry
Austria
ContactUsLinkComponent
 
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit
  CAYM700260-96
 :  
undefined
Anti-Ovalbumin IgG1 (Mouse) ELISA Kit
  CAYM700260-96
 :  

 

  • Pk:
    1
  • Assay Type:
    Sandwich
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Description:
    Anti-ovalbumin IgG1 (mouse) ELISA kitbuffer, substrate, cofactor, initiator, positive control, Plate, and cover Sheet, 96 wells
  • Environmentally Preferable:

 

 

For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.

  • Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
  • Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
  • Assay Range: 1.56 – 200 ng/ml with an LLOQ of 1.56 ng/ml
  • Rapid assay; get results in under 4 hours
  • LLOD: 0.006 U/ml

Cayman's Ovalbumin IgG1 (mouse) ELISA Kit is an immunometric (i.e.,'sandwich') assay which can be used to measure anti-ovalbumin of the IgG1 isotype in mouse plasma and serum without prior sample purification.