Order Entry
Austria
ContactUsLinkComponent
 
Chromata ChIP Histone H3 [Monomethyl Lys9] Kit
  NOVUNBP1-71713
 :  
undefined
Chromata ChIP Histone H3 [Monomethyl Lys9] Kit
  NOVUNBP1-71713
 :  

 

  • Pk:
    25 Tests
  • Description:
    Chromata ChIP Histone H3 [Monomethyl Lys9] kit
  • ChIP:
    Yes
  • Antigen symbol:
    Chromata ChIP Histone H3
  • Product type:
    KIT

 

 

Chromata ChIP Histone H3 [Monomethyl Lys9] Kit

Tested Applications: Chromatin Immunoprecipitation.